missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles

Product Code. 10524408
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
1 L
2 L
5 L
Quantity:
14 x 5 Bottles
20 Bottles
20 x 5 Bottles
35 Bottles
4 x 5 Bottles
6 Bottles
7 x 5 Bottles
70 Bottles
Unit Size:
Pack of 100
Pack of 20
Pack of 35
Pack of 5
Pack of 6
Pack of 70
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity Unit Size
10524408 2 L 4 x 5 Bottles Pack of 20
11319694 125 mL 20 x 5 Bottles Pack of 5
10227747 125 mL 20 x 5 Bottles Pack of 100
10791310 500 mL 14 x 5 Bottles Pack of 70
10237747 500 mL 70 Bottles Pack of 70
12059987 1 L 7 x 5 Bottles Pack of 35
10329587 1 L 35 Bottles Pack of 35
10297887 2 L 20 Bottles Pack of 20
10440568 5 L 6 Bottles Pack of 6
10553298 5 L 6 Bottles Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10524408 Supplier Thermo Scientific™ Supplier No. 323042
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media.

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.

  • Durable, crystal clear PETG for sample integrity
  • Closures with polyethylene foam core liners for leakproof† assurance
  • Custom options available for fluid transfer applications
  • Supported by extensive validation documentation
  • Suitable for storage in conditions from -40° to +70°C

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 L
Color Translucent Blue
Diameter (English) 4.57 in.
Diameter (English) Inner Neck 1.4 in.
Diameter (Metric) Inner Neck 36 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Material PET
No. per Case 20/Cs.
No. per Pack 5/Pk.
Quantity 4 x 5 Bottles
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 265 mm
Height (English) 10.4 in.
Closure Size 48 mm
Diameter (Metric) 116 mm
Autoclavable Not Autoclavable
Workflow Step Upstream process solution storage. Upstream Validation
Capacity (English) 67.63 oz.
Shape Square
Certifications/Compliance USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315
Disposable Single-use
Product Type Square Bottle
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.