missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack
GREENER_CHOICEBIOPHARMA

Product Code. 10150033
Change view
Click to view available options
Capacity (Metric):
3.4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
Unit Size:
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Neck Finish unitSize
10150033 8 mL 20-415 mm Pack of 2000
30117591 3.4 mL 13-415 mm Pack of 2000
10181183 15 mL 20-415 mm Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10150033 Supplier Thermo Scientific™ Supplier No. 3620069025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Safely store and handle test kits and many small volume applications with these PPCO diagnostic bottles, for use in sample storage and shipping small volume reagents.

Safely store and handle test kits and many small volume applications with Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Natural translucent PPCO provides excellent chemical resistance
  • Autoclavable
  • Leakproof† when assembled with closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. Packed 2000 per case. Order colored closures separately (Cat. No. 362150).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.25 oz., 8 mL
Capacity (Metric) 8 mL
Capacity (English) 0.25 oz.
Color Translucent
Diameter (Metric) Inner Neck 14 mm
Material PPCO
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Height (Metric) 45 mm
Product Type Sample Container
Width (English) 0.98 in.
Height (English) 1.77 in.
Diameter (Metric) 25 mm
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.