Learn More
Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack
Safely store and ship small-volume samples and valuable reagents with these PPCO diagnostic bottles, for use in test kits and many small-volume applications.
Brand: Thermo Scientific™ 322006-9050
Code : A0
Additional Details : Weight : 12.50000kg
Description
Safely store and ship small-volume samples and valuable reagents. Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closures are used in test kits and many small-volume applications. These autoclavable and durable polypropylene copolymer bottles are tray packed with closure for easy handling and feature uniform walls resistant to splitting or puncturing.
- Nonsterile
- Natural translucent PPCO with Natural PP Closure
- Durable, uniform walls are resistant to splitting or puncturing
- Bottles are packed in rigid trays allowing for easy handling
- Autoclavable
- Leakproof†
Ordering Information:
Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.Specifications
58 mm | |
Screw Cap | |
Polypropylene | |
25 mm | |
0.5 oz., 15 mL | |
0.5 oz. | |
Natural polypropylene w/PP Closures; Non-sterile; 15mL | |
PPCO | |
150 | |
20-415 mm | |
Yes | |
No | |
Round |
0.98 in. | |
20-415 mm | |
Diagnostic Bottle | |
2.28 in. | |
15 mL | |
Translucent | |
13 mm | |
1500 | |
Non-sterile | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
Autoclavable | |
Narrow |
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.