missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
GREENER_CHOICE

Product Code. 10274401
Click to view available options
Thread Size (Metric):
20 mm
24 mm
28 mm
38 mm
43 mm
53 mm
63 mm
Product Type:
Replacement Cap
Screw Cap
Spout Cap
Stem and Draw Tube
Color:
Amber
Natural
White
Unit Size:
Pack of 12
Pack of 2
Pack of 25
This item is not returnable. View return policy

Product Code. 10274401

Brand: Thermo Scientific™ 7121500430

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.

Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.

  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Description Bottle Replacement Closures
Material Polypropylene
No. per Case 12
Thread Size (Metric) 43 mm
Mouth Wide
Shape Round
Product Type Replacement Cap
For Use With (Equipment) Nalgene narrow-mouth and wide-mouth bottles

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.