missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures

Product Code. 10010681
Change view
Click to view available options
Thread Size (Metric):
20 mm
24 mm
28 mm
38 mm
43 mm
53 mm
63 mm
Product Type:
Replacement Cap
Screw Cap
Spout Cap
Stem and Draw Tube
Color:
Amber
Natural
White
Unit Size:
Pack of 12
Pack of 2
Pack of 25
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Product Type Color Unit Size
10010681 38 mm Replacement Cap Natural Pack of 12
10586551 20 mm Replacement Cap White Pack of 12
10264401 24 mm Replacement Cap Natural Pack of 12
10020661 28 mm Replacement Cap Natural Pack of 12
11334595 28 mm Stem and Draw Tube Natural Pack of 2
10603452 38 mm Replacement Cap White Pack of 12
10799362 38 mm Screw Cap Natural Pack of 12
10274401 43 mm Replacement Cap Natural Pack of 12
10162491 53 mm Replacement Cap Amber Pack of 12
10000721 53 mm Replacement Cap Natural Pack of 2
10050681 53 mm Replacement Cap Natural Pack of 12
10365941 63 mm Replacement Cap Natural Pack of 12
12716677 20 mm Spout Cap Natural Pack of 25
10644212 24 mm Spout Cap Natural Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 10010681 Supplier Thermo Scientific™ Supplier No. 7121600384
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.

Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.

  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Description Bottle Replacement Closures
Material Polypropylene
No. per Case 12
Thread Size (Metric) 38 mm
Mouth Narrow
For Use With (Equipment) Nalgene narrow-mouth and wide-mouth bottles
Shape Round
Product Type Replacement Cap

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.