missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack
GREENER_CHOICEBIOPHARMA

Product Code. 10099252
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-415 mm
24-415 mm
28-415 mm
38-430 mm
Unit Size:
Pack of 1000
Pack of 125
Pack of 250
Pack of 50
Pack of 500

Product Code. 10099252

Brand: Thermo Scientific™ 3120890002

Only null left
Add to basket
Edge
Add to basket

Store, ship and package specialty chemicals, diagnostics, biologicals, reagents, adhesives and veterinary pharmaceuticals with these narrow-mouth HDPE bottles.

Safely store, ship and package liquid reagents using Thermo Scientific™ Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure, an ideal choice for specialty chemicals, diagnostics, biologicals, reagents, adhesives and veterinary pharmaceuticals. The injection blow-molded high-density polyethylene bottles feature a tough plastic construction with uniform wall thickness and strength. Polypropylene closures are included for leakproof† sealing.

Features:

  • Made from high-purity, food-grade resins
  • Recommended for storage and shipment of liquid reagents

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Translucent
Description Narrow-Mouth Packaging Bottles
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 1000
No. per Pack 1000
Closure Color Natural, translucent
Neck Finish 20 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 86 mm
Product Type Packer Bottle
Width (English) 1.52 in.
Height (English) 3.38 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 39 mm
Closure Type Screw (Bulk Packed, Unassembled)
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.