Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays
Easily handle these narrow-mouth HDPE bottles that are ready to feed into filling lines for various reagents, and conveniently packaged in shrink-wrap trays.
£452.62 - £847.58
Specifications
Color | Natural |
---|---|
Description | Narrow-Mouth Packaging Bottles with Closures |
Material | HDPE |
Closure Color | Natural |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10581455
|
Thermo Scientific™
322089-0001 |
30 mL | 20-415 mm |
£717.00
Pack of 1050 |
In Stock
Log in to see stock. |
||||
10435893
|
Thermo Scientific™
322089-0002 |
60 mL | 20-415 mm |
£847.58
Pack of 1050 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
10304243
|
Thermo Scientific™
322089-0008 |
250 mL | 24-415 mm |
£452.62
Pack of 300 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
Description
Easily handle Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles, conveniently packaged in rigid, lint-free shrink-wrap trays and ready to feed into filling lines for various reagents. High density polyethylene bottles include polypropylene closures and rigid, lint-free trays.
Features:- Excellent impact and chemical resistance
- Include opaque amber polypropylene closures
- Translucent amber bottles provide light protection with the ability to see liquid level
- Leakproof† when used with Nalgene closures
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
Spécification
Natural | |
HDPE | |
Yes | |
Round | |
Polypropylene | |
No |
Narrow-Mouth Packaging Bottles with Closures | |
Natural | |
Narrow | |
Narrow Mouth Bottle | |
Screw (Bulk Packed, Unassembled) |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.