missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles, 60mL

Catalog Number 15538199
Click to view available options
Capacity: 2 oz. (60mL)
This item is not returnable. View return policy
This item is not returnable. View return policy

Store, ship and package light-sensitive liquids using these narrow-mouth, amber HDPE bottles that offer excellent chemical resistance to most acids, bases and alcohols.

Store, ship and package light-sensitive liquids using Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles. Featuring excellent chemical resistance to most acids, bases and alcohols, these high-density polyethylene bottles are guaranteed leakproof†. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:

  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Versatile bottles where exposure to light needs to be restricted
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaiminate your reagents

Includes:

Amber polypropylene screw caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

TRUSTED_SUSTAINABILITY
Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Opaque Amber
Description Lab Quality Amber Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Type Lab Quality Bottle
Closure Color Amber
Neck Finish 20 mm
Certifications/Compliance Meet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
Closure Liner Linerless
Closure Included Yes
Closure Size 20 mm
Closure Material Polypropylene
Closure Type Screw Cap
Diameter (Metric) 37 mm
Graduated No
Height (English) 3.36 in.
Height (Metric) 85 mm
Mouth Narrow
Shape Round
Through the Closure Dispensing No
Width (English) 1.52 in.
Show More Show Less

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'