Learn More
Thermo Scientific™ Nalgene™ Microcentrifuge Tube Storage Boxes, 0.5mL tubes
Safely store samples using these microcentrifuge tube storage boxes.
Brand:  Thermo Scientific™ 916912
Product Code. 11310495
  • £220.00 / Pack of 672
Only null left
In Stock
Add to basket
Description
Description
Safely store samples using Thermo Scientific™ Nalgene™ Microcentrifuge Tube Storage Boxes. High-temperature polyurethane foam insert securely holds 64 or 81 microcentrifuge tubes sized 0.2, 0.5 or 1.5mL.
  • Bottom constructed of white polycarbonate
  • Transparent polycarbonate lid has printed grid for inventory purposes
  • Store from -211° to +250°F (-135° to +121°C)
  • Lid accepts marker for box and tube identification and is keyed to orient front and back of box
  • L × W × H: 5.25 × 5.25 × 2″ (13 × 13 × 5cm)
Note:
Not for use in liquid phase of liquid nitrogen.