missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack

Catalog Number p-4521599
Click to view available options
Capacity (Metric):
40 μL
50 μL

Carry out pharmaceutical and biotech applications with these LDPE dispensing tips for dropper bottles, which feature precise drip control.

Carry out pharmaceutical and biotech applications with Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles. Tips feature precise drip control and can be used with Nalgene LDPE Dropper bottles and Colored PP Closures.

Features:
  • Low density polyethylene
  • Offered in two drop sizes (40μL or 50μL)
  • Fits Nalgene Dropper Bottles Series 312750 and 312751
  • Use with Nalgene dropper closures for color coding your reagents

Ordering Information:

Dropper Bottles Dispensing Tips are bulk packed 2000 per case. Order Nalgene dropper bottles (Cat. Nos. 312750, 312751) and colored closure (Cat. No. 312760) separately.

TRUSTED_SUSTAINABILITY
Diameter (Metric) 11.22 mm
For Use With (Equipment) Nalgene Controlled Dropper
Height (English) 0.65 in.
Height (Metric) 16.5 mm
Width (English) 0.39 in.
Product Type Tips for Dropper Bottle
Capacity (Metric) 40 μL
Capacity (English) 1.35 oz.
Color Clear
Material LDPE
No. per Case 2000
No. per Pack 2000
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle dispensing tips meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Drop Size 40/50 μL
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'