missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
GREENER_CHOICE

Product Code. 10481371
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10481371

Brand: Thermo Scientific™ 21230010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.

Collect, store and transport dry chemicals and environmental samples with Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure. Molded-in handgrips provide greater convenience and safety when pouring.

  • Useful for collecting, storing and transporting dry chemicals, environmental samples, reagents and distilled water
  • Good for freezing to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols
  • Square shape saves space, handles easily
  • Wide mouth gives easy access
  • Polypropylene cap included
  • Not liquid leakproof
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 4 L
Capacity (English) 1 gal.
Color Translucent
Description Large Wide-Mouth Square Bottle
Diameter (English) 5.6 in.
Diameter (English) Inner Neck 3.5 in.
Diameter (Metric) Inner Neck 89 mm
Material HDPE
No. per Pack 1
Sterility Non-sterile
Closure Color White
Certifications/Compliance USP Class VI; FDA - 21 CFR
Closure Included Yes
Graduated No
Mouth Wide
Shape Square
Height (Metric) 292 mm
Product Type Wide Mouth Bottle (Large)
Width (English) 5.66 in.
Height (English) 11.5 in.
Closure Material Polypropylene
Closure Size 100 mm
Diameter (Metric) 142 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.