missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Narrow-Mouth Amber Bottles
GREENER_CHOICE

Product Code. 10420913
Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Closure Size:
38-430 mm
53B
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10420913

Brand: Thermo Scientific™ 22040010

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protect light-sensitive liquids with these amber bottles that reduce UV light transmissions and feature excellent chemical resistance to most acids, bases and alcohols.

Protect light-sensitive liquids with Thermo Scientific™ Nalgene™ Large Amber Narrow-Mouth Bottles, which reduce UV light transmissions. Useful for storing chemicals and standards, as well as for collecting and storing distilled water, these bottles display excellent chemical resistance to most acids, bases and alcohols.

Features:

  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meet the U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Autoclavable/Leakproof†

Note:

Completely disengage threads or remove cap before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 gal., 4 L
Capacity (Metric) 4 L
Capacity (English) 1 gal.
Color Amber
Description Amber Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 25 mm
Material PPCO
No. per Case 6
No. per Pack 1
Sterility Non-sterile
Closure Color Amber
Neck Finish 38 mm
Certifications/Compliance Comply with U.S. Pharmacopeia XXIII requirements for maximum allowable light transmission
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 333 mm
Product Type Narrow Mouth Bottle
Height (English) 13.1 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 152 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.