missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE, Jerry Can with Tethered Polypropylene Closure (Double Port)

Product Code. 15911491
Click to view available options
:
With double port; 3.5 gal. (13L)
This item is not returnable. View return policy

Product Code. 15911491

Brand: Thermo Scientific™ 22439013

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Use With: Ideal as a reservoir in automated systems; Closure Material: Polypropylene; Color: White

Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure, featuring a space-saving design, molded-in handle and recessed grip at the bottom to provide better control when pouring. The double-ported 2243-9013 is ideal as a reservoir in automated systems. The 53B closure can be plumbed with input/output tubing while the 38mm closure is used for refilling or emptying the reservoir.

Key Features:
  • Jerrican is ideal as a reservoir in automated systems like the PRISM™ 3700 DAN Analyzer
  • Molded-in graduations in 1 gal. and 5L increments allow for ease in dispensing
  • Molded-in handle provides a sturdy grip when carrying heavy contents
  • Integral shoulder handle and molded bottom grip make lifting comfortable
  • Wide stance and low center of gravity add stability
  • Heavy rugged design with good chemical resistance allows for a wide variety of solutions
  • Space-saving rectangular shape permits efficient storage

Ordering Information:

  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • Custom tubing sets can be designed at the Thermo Scientific Custom Tubing Set Configurator, www.thermoscientific.com/tubingconfigurator
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 3.5 gal.
Capacity (Metric) 13 L
Certifications/Compliance USP Class VI Compliant/FDA21 CFR 177.1520 Compliant
Closure Color White
Closure Material Polypropylene
Closure Size 53B
Closure Type Tethered
Color White
Description Jerrican w/double port
Diameter (English) Inner Neck 1.5 in.
Diameter (Metric) Inner Neck 38 mm
For Use With (Application) Ideal as a reservoir in automated systems
Material HDPE
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Jerry Can
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.