missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
GREENER_CHOICEBIOPHARMA

Product Code. 10190363
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
Unit Size:
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Neck Finish unitSize
10190363 4 mL 13-415 mm Pack of 2000
10797443 8 mL 20-415 mm Pack of 2000
10069252 15 mL 20-415 mm Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190363 Supplier Thermo Scientific™ Supplier No. 3620089125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with these HDPE diagnostic bottles without closure.

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure. Safe and leakproof† when used with Nalgene closures, these bottles can also be used in test kits and many other small-volume applications. Bottles are made of opaque white, high-density polyethylene and are packed without closures. They can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof† when assembled with Nalgene closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. 2000 per case. Order colored closures separately (Cat. No. 362150).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.125 oz., 4 mL
Capacity (Metric) 4 mL
Capacity (English) 0.125 oz.
Color Opaque White
Diameter (Metric) Inner Neck 8 mm
Material HDPE
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 13-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Included No
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 41 mm
Product Type Sample Container
Width (English) 0.62 in.
Height (English) 1.61 in.
Closure Material Polypropylene
Closure Size 13-415 mm
Diameter (Metric) 16 mm
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.