missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack
GREENER_CHOICE

Product Code. 10562425
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
Unit Size:
Pack of 1328
Pack of 1500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Neck Finish unitSize
10562425 4 mL 13-415 mm Pack of 1328
10252913 8 mL 20-415 mm Pack of 1500
10019022 15 mL 20-415 mm Pack of 1500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10562425 Supplier Thermo Scientific™ Supplier No. 3220029125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform many small-volume applications with these safe diagnostic bottles that are leakproof when used with Nalgene closures. Ideal for use in test kits, sample storage and reagent shipping.

Perform many small-volume applications with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure. These bottles are ideal for use in test kits, sample storage and shipping of small-volume, valuable reagents. They come packed in rigid trays for easy handling.

Natural High Density Polyethylene Bottles:
  • Nonsterile
  • Translucent
  • Natural polypropylene (PP) closure for excellent chemical resistance
  • Durable, uniform walls that are resistant to splitting or puncturing
  • Leakproof†

Ordering Information:

Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.125 oz., 4 mL
Capacity (Metric) 4 mL
Capacity (English) 0.125 oz.
Color Translucent
Diameter (Metric) Inner Neck 8 mm
Material HDPE
No. per Case 1328
No. per Pack 332
Sterility Non-sterile
Neck Finish 13-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Height (Metric) 41 mm
Product Type Sample Container
Width (English) 0.62 in.
Height (English) 1.61 in.
Diameter (Metric) 16 mm
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.