Learn More
Thermo Scientific™ CryoTube™ Holders
Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.
Brand:  Thermo Scientific™ 376589
Code : 76
Additional Details : Weight : 2.20000kg
 View more versions of this product
Product Code. 11381665
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.
  • Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
  • Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
Specifications
Specifications
Nunc™ Tube/Vial Containers
PPO
Nunc™
4 x 10
Interlocking
Vial Closure
1/Pk., 15/Cs.