missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles

Catalog Number p-6137224
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Closure Color:
Assorted
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Use for accurate, reliable, repeatable dispensing of various reagents. These dropper bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. An excellent alternative to pipetting devices.

Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.

  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included, choose from white or multi-colored caps
  • Leakproof†
TRUSTED_SUSTAINABILITY
Closure Size 15-415 mm
Closure Type Dropper Control Tip
Through the Closure Dispensing Yes
Product Type Dropping Bottle
Capacity (Metric) 4 mL
Capacity (English) 0.13 oz.
Color White
Description LDPE Dropper Bottle
Includes Dispensing tip and closures
Material LDPE
No. per Case 25
No. per Pack 25
Closure Color White
Certifications/Compliance ISO 13485:2003
Closure Included Yes
Drop Size 40 μL
Shape Round
Show More Show Less

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'