missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles
GREENER_CHOICE

Product Code. 10447452
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Closure Color:
Assorted
White
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
10447452 8 mL White Pack of 25
10739441 4 mL White Pack of 25
10332862 15 mL Assorted Pack of 25
10532093 15 mL White Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10447452 Supplier Thermo Scientific™ Supplier No. 27519025
Curiosity Goes Beyond
Use promo code 27966
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Use for accurate, reliable, repeatable dispensing of various reagents. These dropper bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. An excellent alternative to pipetting devices.

Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.

  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included, choose from white or multi-colored caps
  • Leakproof†
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 8 mL
Color White
Description LDPE Dropper Bottle
Includes Dispensing tip and closures
Material LDPE
No. per Case 25
No. per Pack 25
Closure Color White
Closure Included Yes
Dropper Capacity 8 mL
Drop Size 40 μL
Closure Size 15-415 mm
Closure Type Dropper Control Tip
Through the Closure Dispensing Yes
Capacity (English) 0.27 oz.
Shape Round
Product Type Dropping Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.