missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip
GREENER_CHOICE

Product Code. p-4521605
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Closure Color:
Assorted
White
missing translation for 'unitSize'
Pack of 25
This item is not returnable. View return policy

Product Code. 10632283

missing translation for 'mfr': Thermo Scientific™ 27529125

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Repeatedly dispense reagents accurately using these dropper bottles, an excellent alternative to pipetting devices.

Repeatedly dispense reagents accurately using Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip. An excellent alternative to pipetting devices, these dropper bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Featuring excellent chemical resistance, these specialty bottles are suitable for most biotech, diagnostic and pharmaceutical applications.

Features:
  • Contact-clear, natural, low-density polyethylene bottle for viewing content
  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included; chose white or multi-colored closures
  • Leakproof†/Transparent

Ordering Information:

This product comes in a case of 25.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 4 mL
Capacity (English) 0.13 oz.
Color Translucent
Material LDPE
No. per Case 25
No. per Pack 25
Closure Color Assorted
Certifications/Compliance ISO 13485:2003
Closure Included Yes
Drop Size 40 μL
Shape Round
Height (Metric) 53.8 mm
Product Type Dropping Bottle
For Use With (Equipment) Nalgene Closures
Width (English) 0.78 in.
Height (English) 2.12 in.
Closure Size 15-415 mm
Through the Closure Dispensing Yes
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato