missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip
GREENER_CHOICE

Product Code. 10080841
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Closure Color:
Assorted
White
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color unitSize
10080841 4 mL White Pack of 25
10632283 4 mL Assorted Pack of 25
10396331 8 mL White Pack of 25
10203202 8 mL Assorted Pack of 25
10778281 15 mL White Pack of 25
10181382 15 mL Assorted Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10080841 Supplier Thermo Scientific™ Supplier No. 27509125
Add to basket
Add to basket

Repeatedly dispense reagents accurately using these dropper bottles, an excellent alternative to pipetting devices.

Repeatedly dispense reagents accurately using Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip. An excellent alternative to pipetting devices, these dropper bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Featuring excellent chemical resistance, these specialty bottles are suitable for most biotech, diagnostic and pharmaceutical applications.

Features:
  • Contact-clear, natural, low-density polyethylene bottle for viewing content
  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included; chose white or multi-colored closures
  • Leakproof†/Transparent

Ordering Information:

This product comes in a case of 25.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 4 mL
Capacity (English) 0.13 oz.
Color Translucent
Material LDPE
No. per Case 25
No. per Pack 25
Closure Color White
Certifications/Compliance ISO 13485:2003
Dropper Capacity 4 mL
Drop Size 40 μL
Shape Round
Height (Metric) 53.8 mm
Product Type Dropping Bottle
Width (English) 0.78 in.
Height (English) 2.12 in.
Closure Size 15-415 mm
Diameter (Metric) 19.7 mm
Through the Closure Dispensing Yes
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.