missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles
GREENER_CHOICE

Product Code. 10780442
Change view
Click to view available options
Closure Color:
Blue
Green
Red
Yellow
Unit Size:
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color unitSize
10780442 Red Pack of 6
10055280 Yellow Pack of 6
10732191 Blue Pack of 6
10517131 Green Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10780442 Supplier Thermo Scientific™ Supplier No. 24225500
Only null left
Add to basket
Add to basket

Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.

High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

  • Color-coded top allows you to coordinate reagents with industry storage codes.
  • Tip can be cut back to provide increased flow.
  • Tight fit of draw tube and closure prevents separation when squeezing bottle.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Natural
Description With stem and draw tube
Material LDPE
No. per Case 24
No. per Pack 6
Closure Color Red
Closure Included Yes
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 224 mm
Product Type Color-coded Wash Bottle
Height (English) 8.8 in.
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 72.9 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.