missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners

Product Code. 10050063
Change view
Click to view available options
Capacity (English):
5 gal.
7.5 gal.
10 gal.
15 gal.
30 gal.
55 gal.
80 gal.
100 gal.
150 gal.
200 gal.
Unit Size:
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Unit Size
10050063 10 gal. Pack of 10
10511315 5 gal. Pack of 10
10141133 7.5 gal. Pack of 10
10424423 15 gal. Pack of 10
10561695 30 gal. Pack of 10
10767104 55 gal. Pack of 10
10069632 80 gal. Pack of 10
10405183 100 gal. Pack of 10
10079632 150 gal. Pack of 10
10243243 200 gal. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10050063 Supplier Thermo Scientific™ Supplier No. 3430500010
Spotlight Offers
Use promo code 28038
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.

Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.

  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Description Gamma Irradiated: 10 gal. (38L)
Type Tank Liner
Material Polyethylene
Capacity (English) 10 gal.
Capacity (Metric) 38 L
For Use With (Equipment) Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case 10
No. per Pack 10

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.