missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Carboy Replacement Screw Closures and Gaskets

Product Code. 10700333
Change view
Click to view available options
Neck Finish:
100-400
120-400
53B
83B
Material:
HDPE
Polypropylene
Silicone
TPE
Product Type:
Screw Cap
Screw Cap with Strap
Screw Closure and Gasket
Sealing Gasket
Unit Size:
Each
Pack of 10
Pack of 12
Pack of 2
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Material Product Type Unit Size
10700333 83B TPE Sealing Gasket Pack of 5
10244451 100-400 Polypropylene Screw Cap Pack of 12
10632102 120-400 Polypropylene Screw Cap Pack of 12
10789031 53B HDPE Screw Cap with Strap Pack of 10
10652292 53B Polypropylene Screw Cap Pack of 12
10254451 53B HDPE Screw Closure and Gasket Pack of 12
10376081 53B TPE Sealing Gasket Pack of 12
10525571 83B Polypropylene Screw Cap Pack of 2
10152351 83B HDPE Screw Cap Pack of 2
10214451 83B Silicone Screw Cap Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10700333 Supplier Thermo Scientific™ Supplier No. 7121621830
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service.

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

  • Made to fit Nalgene Carboys
  • Leakproof† when used with the Nalgene bottles with the matching finish
  • TRUSTED_SUSTAINABILITY

    Specifications

    Color White
    Description Sealing TPE (thermoplastic elastomer) Gasket
    Material TPE
    No. per Case 5
    No. per Pack 5
    Sterility Non-sterile
    Neck Finish 83B
    For Use With (Equipment) Nalgene Large Carboys and Bottles
    Closure Size 83B
    Autoclavable Autoclavable
    Disposable Single-use
    Product Type Sealing Gasket
    Show More Show Less

    † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.