missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NALCN Monoclonal Antibody (N187/7), PerCP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545574
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45574 was sufficient for detection of NALCN in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 200kDa. This antibody was formerly sold as clone S187-7.
Nalcn is a voltage-independent, cation-nonselective channel which is permeable to sodium, potassium and calcium ions. Nalcn regulates the resting membrane potential and controls neuronal excitability (PubMed:17448995). Neuropeptides such as neurotensin and substance P (SP) stimulate the firing of action potentials by activating NALCN through a SRC family kinases-dependent pathway. In addition to its baseline activity, Nalcn activity is enhanced/modulated by several GPCRs, is required for normal respiratory rhythm and neonatal survival, is involved in systemic osmoregulation by controlling the serum sodium concentration. Nalcn is partly responsible for the substance P-induced depolarization and regulation of the intestinal pace-making activity in the interstitial cells of Cajal. Further, Nalcn plays a critical role in both maintenance of spontaneous firing of substantia nigra pars reticulata (SNr) neurons and physiological modulation of SNr neuron excitability. Diseases associated with NALCN include Congenital contractures of the limbs and face, Hypotonia, Developmental Delay and Hypotonia, Infantile Psychomotor Retardation And Characteristic Facies 1.
Specifications
| NALCN | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q6Q760, Q8BXR5, Q8IZF0 | |
| NALCN | |
| Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| N187/7 | |
| PerCP | |
| NALCN | |
| A530023G15Rik; AI849508; bA430M15.1; brain voltage-gated cation channel; CanIon; CLIFAHDD; FLJ23913; FLJ44659; FLJ44764; Four domain-type voltage-gated ion channel alpha-1 subunit; four repeat voltage-gated ion channel; IHPRF; IHPRF1; INNFD; MGC74524; Nalcn; Nca; Rb21; Rb21-channel; Sodium leak channel non-selective protein; sodium leak channel, non selective; sodium leak channel, non-selective; Vgcnl1; voltage gated channel like 1; voltage gated channel-like protein 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 259232, 266760, 338370 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction