missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Naked2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561979
This item is not returnable.
View return policy
Description
Immunogen sequence: GHKRYRQKGR EGHSPLKAPH AQPATVEHEV VRDLPPTPAG EGYAVPVIQR HE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.
Cell autonomous antagonist of the canonical Wnt signaling pathway. May activate a second Wnt signaling pathway that controls planar cell polarity. Required for processing of TGFA and for targeting of TGFA to the basolateral membrane of polarized epithelial cells.
Specifications
| Naked2 | |
| Polyclonal | |
| Unconjugated | |
| Nkd2 | |
| 2210403L10Rik; AW212591; Dvl-binding protein NKD2; hNkd2; mNkd2; naked cuticle 1 homolog; naked cuticle 2 homolog; naked cuticle 2 homolog (Drosophila); naked cuticle homolog 2; naked cuticle homolog 2 (Drosophila); naked cuticle-2; Naked2; naked-2; Nkd2; Protein naked cuticle homolog 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969F2 | |
| Nkd2 | |
| Recombinant protein corresponding to Human Naked2. Recombinant protein control fragment (Product #RP-96881). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction