missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Naked2 Monoclonal Antibody (S.902.7)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514827
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Cell autonomous antagonist of the canonical Wnt signaling pathway. May activate a second Wnt signaling pathway that controls planar cell polarity. Required for processing of TGFA and for targeting of TGFA to the basolateral membrane of polarized epithelial cells.
Specifications
| Naked2 | |
| Monoclonal | |
| Unconjugated | |
| Nkd2 | |
| 2210403L10Rik; AW212591; Dvl-binding protein NKD2; hNkd2; mNkd2; naked cuticle 1 homolog; naked cuticle 2 homolog; naked cuticle 2 homolog (Drosophila); naked cuticle homolog 2; naked cuticle homolog 2 (Drosophila); naked cuticle-2; Naked2; naked-2; Nkd2; Protein naked cuticle homolog 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 85409 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| S.902.7 | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q969F2 | |
| Nkd2 | |
| Synthetic peptide corresponding to residues surrounding Ala217 of human Naked2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction