missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Naja kaouthia Cobra Venom Factor Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117705
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Complement-activating protein in cobra venom. It is a structural and functional analog of complement component C3b, the activated form of C3. It binds factor B (CFB), which is subsequently cleaved by factor D (CFD) to form the bimolecular complex CVF/Bb. CVF/Bb is a C3/C5 convertase that cleaves both complement components C3 and C5. Structurally, it resembles the C3b degradation product C3c, which is not able to form a C3/C5 convertase. Unlike C3b/Bb, CVF/Bb is a stable complex and completely resistant to the actions of complement regulatory factors H (CFH) and I (CFI). Therefore, CVF continuously activates complement resulting in the depletion of complement activity.
Specifications
| Naja kaouthia Cobra Venom Factor | |
| Polyclonal | |
| Unconjugated | |
| CVF | |
| Recombinant Naja kaouthia Cobra venom factor protein (733-984aa). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Reptile | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction