missing translation for 'onlineSavingsMsg'
Learn More

NAIP Antibody [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30735205 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735205 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735205 Supplier Novus Biologicals Supplier No. AF7608J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

NAIP Polyclonal antibody specifically detects NAIP in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAIP
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias baculoviral IAP repeat-containing 1, BIRC1baculoviral IAP repeat-containing protein 1, FLJ18088, FLJ42520, FLJ58811, Neuronal apoptosis inhibitory protein, NLR family, apoptosis inhibitory protein, NLR family, BIR domain containing 1, NLRB1, nucleotide-binding oligomerization domain, leucine rich repeat and BIR domaincontaining 1, psi neuronal apoptosis inhibitory protein, psiNAIP
Host Species Sheep
Immunogen E. coli-derived recombinant mouse NAIP, Lys783-Ser856, Accession # Q9QWK5
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4671
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.