missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAIP Recombinant Rabbit Monoclonal Antibody (13E6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550166
This item is not returnable.
View return policy
Description
NAIP Recombinant Monoclonal Antibody for IHC (P), ELISA
Neuronal apoptosis inhibitor protein (NAIP) was the first human inhibitor of apoptosis protein (IAP) identified and was discovered by its association with the neurodegenerative disorder spinal muscular atrophy. Members of the IAP family contain one to three copies of an approximately 70 amino acid motif termed baculovirus IAP repeat (BIR); these BIRs promote protein-protein interactions with various caspases such as caspase-3, -7, and -9 as well as members of the TRAF family of signal molecules (reviewed in 3). Unlike other IAPs however, NAIP requires ATP to bind caspase-9 and is not inhibited by the IAP-inhibiting molecule Smac/DIABLO, suggesting that NAIP is unique among the IAPs in its regulation of its activity. Finally, although only one human NAIP protein has been identified, other shorter NAIP mRNA transcripts have been reported.
Specifications
| NAIP | |
| Recombinant Monoclonal | |
| 1.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13075 | |
| NAIP | |
| A synthesized peptide derived from Human NAIP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 13E6 | |
| Unconjugated | |
| NAIP | |
| AV364616; baculoviral IAP repeat-containing 1a; baculoviral IAP repeat-containing protein 1; Baculoviral IAP repeat-containing protein 1a; BIRC1; Birc1a; D13Lsd1; FLJ18088; FLJ42520; FLJ58811; Naip; Naip1; Naip-rs1; Neuronal apoptosis inhibitory protein; neuronal apoptosis inhibitory protein 1; NLR family apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein 1; NLRB1; nucleotide-binding oligomerization domain, leucine rich repeat and BIR domain containing 1; psi neuronal apoptosis inhibitory protein; psiNAIP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4671 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction