missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAIF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68824
This item is not returnable.
View return policy
Description
NAIF1 Polyclonal antibody specifically detects NAIF1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| NAIF1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| bA379C10.2, C9orf90RP11-379C10.2, chromosome 9 open reading frame 90, DKFZp762G199, nuclear apoptosis inducing factor 1, nuclear apoptosis-inducing factor 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EQQNDLLQMIRRSQEVQACAQERQAQAMEGTQAALSVLIQVLRPMIKDFRRYLQSNTANPAPASDPGQVAQNGQPDSIIQ | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication | |
| 203245 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction