missing translation for 'onlineSavingsMsg'
Learn More

NAI-N3, MedChemExpress

Product Code. 16406497
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16406497 5 mg 5mg
16463768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16406497 Supplier Medchem Express Supplier No. HY103006
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NAI-N3 is a RNA acylation reagent that enables RNA purification. NAI-N3 is a dual-function SHAPE (selective 2-hydroxyl acylation and profiling experiment) probe (RNA structure probe and enrichment).

NAI-N3 is a RNA acylation reagent that enables RNA purification. NAI-N3 is a dual-function SHAPE (selective 2-hydroxyl acylation and profiling experiment) probe (RNA structure probe and enrichment).

Specifications

Chemical Name or Material NAI-N3
CAS 1612756-29-2
Color White
Purity Grade Notes Research
Molecular Formula C10H8N6O
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (1095.48 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=CC=CN=C1CN=[N+]=[N-])N2C=CN=C2
Molecular Weight (g/mol) 228.21
Formula Weight 228.21
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.