missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NAI
GREENER_CHOICE

Product Code. 18729706
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18729706 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18729706 Supplier Tocris Bioscience™ Supplier No. 7004/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent for RNA SHAPE-MaP experiments in vivo; cell permeable

NAI is a reagent for RNA SHAPE-MaP experiments in live cells. Acylates the 2'-OH in RNA, labels single stranded or flexible regions more than folded regions (SHAPE methodology). Enables analysis of RNA secondary structures at single nucleotide resolution. Exhibits greater signals with lower background than 1M7 for in vivo measurement of the RNA structurome. Also more stable than 1M7 in water. β-mercaptoethanol terminates reaction. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1 H-Imidazol-1-yl(2-methyl-3-pyridinyl)methanone
CAS 1055970-47-2
Quantity 10 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C10H9N3O
Formula Weight 2424
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.