missing translation for 'onlineSavingsMsg'
Learn More

Nagilactone B, MedChemExpress

Product Code. 16422599
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422599 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422599 Supplier Medchem Express Supplier No. HYN3216

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nagilactone B is a liver X receptor (LXR) agonist.

Nagilactone B is a liver X receptor (LXR) agonist.

Specifications

Chemical Name or Material Nagilactone B
CAS 19891-51-1
Color White
Purity Grade Notes Research
Molecular Formula C19H24O7
Quantity 1 mg
SMILES C[C@@]1(C2=C3)[C@@]([C@@]4(C[C@@H](O)[C@H]1O)C)([H])[C@@](OC4=O)([H])[C@H](O)C2=C(C(C)C)OC3=O
Molecular Weight (g/mol) 364.39
Formula Weight 364.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.