missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAG Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18441AF594
This item is not returnable.
View return policy
Description
NAG Polyclonal antibody specifically detects NAG in Human samples. It is validated for Western Blot
Specifications
| NAG | |
| Polyclonal | |
| Western Blot | |
| DKFZp586G1219, EC 1.1.1.94, EC 1.2.1.59, FLJ40407, NAG/BC035112 fusion, NAG/FAM49A fusion, NAGneuroblastoma-amplified protein, neuroblastoma amplified sequence, Neuroblastoma-amplified gene protein, neuroblastoma-amplified sequence | |
| Recognizes region between residues Between Between 350 and 400.. | |
| 0.1 mL | |
| Cell Biology | |
| 51594 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction