missing translation for 'onlineSavingsMsg'
Learn More

Naftifine hydrochloride, MedChemExpress

Product Code. 16400978
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16400978

Brand: Medchem Express HYB0518A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Naftifine hydrochloride is an antibiotic. Naftifine hydrochloride has antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. Naftifine hydrochloride can be used for the research of superficial dermatomycoses inhibition.

Naftifine hydrochloride is an antibiotic. Naftifine hydrochloride has antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. Naftifine hydrochloride can be used for the research of superficial dermatomycoses inhibition.

Specifications

Chemical Name or Material Naftifine hydrochloride
Physical Form Solid
CAS 65473-14-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C21H22ClN
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (308.78 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (77.19 mM; Need ultrasonic)
SMILES CN(C/C=C/C1=CC=CC=C1)CC2=CC=CC3=CC=CC=C23.Cl
Molecular Weight (g/mol) 323.86
Formula Weight 323.86
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.