missing translation for 'onlineSavingsMsg'
Learn More

Nafion∣r NRE-212 membrane, 0.05mm thick, ∣>0.92 meq/g exchange capacity

Product Code. 15496685
Change view
Click to view available options
Quantity:
15 x 15 cm
30 x 30 cm
60 x 60 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496685 15 x 15 cm Each
15486685 30 x 30 cm Each
15476685 60 x 60 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15496685 Supplier Thermo Scientific Alfa Aesar Supplier No. 045036.VA
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Has a wide variety of commercial uses. It is an acidic hydrogen ion form and superacid catalysts useful in a wide variety of synthetic applications. | CAS: 31175-20-9 | C9HF17O5S | 544.135 g/mol

Nafion™ NRE-212 membrane is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. It is an acidic hydrogen ion form and superacid catalysts useful in a wide variety of synthetic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31175-20-9
Molecular Formula C9HF17O5S
Molecular Weight (g/mol) 544.135
MDL Number MFCD00132790
InChI Key FOYUGSIADQEOEK-UHFFFAOYSA-N
Synonym nafion, nafion nr 50 beads 10-35 mesh, perfluoro 2-2-sulfonylethoxy propyl vinyl ether-tetrafluoroethylene copolymer, tetrafluoroethylene-perfluoro 2-2-sulfonylethoxy propyl vinyl ether copolymer, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoroethanesulfonyl fluoride-tetrafl, ethanesulfonic acid, 1,1,2,2-tetrafluoro-2-1,2,2-trifluoro-1-trifluoromethyl-2-1,2,2-trifluorovinyl oxy ethoxy-, pol, ethanesulfonic acid, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer, nafion n-324 membrane, 0.15mm 0.006in thick, teflon fabric reinforced, nafion nre-212 membrane, 0.05mm thick, ≥0.92 meq/g exchange capacity, nafion d-520 dispersion w/w in water and 1-propanol, ≥1.00 meq/g exchange capacity
PubChem CID 61889
IUPAC Name 1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonic acid
SMILES C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)O)(F)F)F)(F)F)F.C(=C(F)F)(F)F

Specifications

Packaging Shrink-wrapped
Quantity 15 x 15 cm
Solubility Information Insoluble in water.
Chemical Name or Material Nafion™ NRE-212 membrane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.