missing translation for 'onlineSavingsMsg'
Learn More

Nafion∣r D-521 dispersion, 5% w/w in water and 1-propanol, ∣>0.92 meq/g exchange capacity

Product Code. 11369777
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
25mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369777 25 mL 25mL
11359777 100 mL 100mL
11366049 500 mL 500mL
11379777 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11369777 Supplier Thermo Scientific Alfa Aesar Supplier No. 042117.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Preparation of thin films and coatings; proton exchange membranes | CAS: 31175-20-9 | C9HF17O5S | 544.135 g/mol

Preparation of thin films and coatings; proton exchange membranes.Nafion™ D-521 dispersion, 5% w/w in water and 1-propanol is used in the fabrication of thin films and coating formulations for fuel cell membranes, catalyst coatings, sensors, and a variety of electrochemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31175-20-9
Molecular Formula C9HF17O5S
Molecular Weight (g/mol) 544.135
MDL Number MFCD00132790
InChI Key FOYUGSIADQEOEK-UHFFFAOYSA-N
Synonym nafion, nafion nr 50 beads 10-35 mesh, perfluoro 2-2-sulfonylethoxy propyl vinyl ether-tetrafluoroethylene copolymer, tetrafluoroethylene-perfluoro 2-2-sulfonylethoxy propyl vinyl ether copolymer, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoroethanesulfonyl fluoride-tetrafl, ethanesulfonic acid, 1,1,2,2-tetrafluoro-2-1,2,2-trifluoro-1-trifluoromethyl-2-1,2,2-trifluorovinyl oxy ethoxy-, pol, ethanesulfonic acid, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer, nafion n-324 membrane, 0.15mm 0.006in thick, teflon fabric reinforced, nafion nre-212 membrane, 0.05mm thick, ≥0.92 meq/g exchange capacity, nafion d-520 dispersion w/w in water and 1-propanol, ≥1.00 meq/g exchange capacity
PubChem CID 61889
IUPAC Name 1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonic acid
SMILES C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)O)(F)F)F)(F)F)F.C(=C(F)F)(F)F

Specifications

Color Colorless to Yellow
Physical Form Liquid
UN Number UN1987
Density 0.93 g/mL
Solubility Information Insoluble in water.
Quantity 25 mL
Flash Point 24°C (75°F)
Concentration or Composition (by Analyte or Components) 5% w/w in water and 1-propanol
Chemical Name or Material Nafion™ D-521 dispersion, ≥0.92meq/g exchange capacity

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.