missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nafamostat mesylate
GREENER_CHOICE

Product Code. 18792684
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18792684

Brand: Tocris Bioscience™ 3081/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serine protease inhibitor, inhibits tryptase

Nafamostat mesylate is a broad spectrum serine protease inhibitor. Displays selectivity for human tryptase when used at relatively low concentrations. Reduces eosinophil infiltration, mast cell activation and airway responsiveness in a murine model of asthma. Inhibits SARS-CoV-2 infection in vitro (EC50 = 22.50 μM).

Tocris products are for biomedical research use only. They are not intended for human or veterinary use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82956-11-4
Molecular Formula C21H25N5O8S2

Specifications

Quantity 10 mg
Formula Weight 539.59
Purity >99%
Molecular Weight (g/mol) 539.58
Chemical Name or Material Nafamostat mesylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.