missing translation for 'onlineSavingsMsg'
Learn More

Nafamostat mesylate, MedChemExpress

Product Code. 16458398
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16458398

Brand: Medchem Express HYB0190A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nafamostat mesylate, a synthetic serine protease inhibitor, is an anticoagulant. Nafamostat mesylate supresses T cell auto-reactivity by decreasing granzyme activity and CTL cytolysis. Nafamostat mesylate blocks activation of SARS-CoV-2..

Nafamostat mesylate, a synthetic serine protease inhibitor, is an anticoagulant. Nafamostat mesylate supresses T cell auto-reactivity by decreasing granzyme activity and CTL cytolysis. Nafamostat mesylate blocks activation of SARS-CoV-2..

Specifications

Chemical Name or Material Nafamostat mesylate
CAS 82956-11-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25N5O8S2
Quantity 10 mg
Synonym FUT-175
Solubility Information DMSO : 50 mg/mL (92.66 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (61.77 mM; Need ultrasonic)
SMILES CS(=O)(O)=O.O=C(OC1=CC=C2C=C(C(N)=N)C=CC2=C1)C3=CC=C(NC(N)=N)C=C3.CS(=O)(O)=O
Molecular Weight (g/mol) 539.58
Formula Weight 539.58
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.