missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557582
This item is not returnable.
View return policy
Description
Immunogen sequence: EIFGPVAHPF YVLRFNSSDH IESKGIKIKE TMYFAPSMKD FTQYIFTEKL KQDKGSDASW KNDQEPPPEA LDFSDDEKEK EAKQRKKSQI QGRK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
RNA-binding protein required for the maturation of box H/ACA snoRNPs complex and ribosome biogenesis. During assembly of the H/ACA snoRNPs complex, it associates with the complex and disappears during maturation of the complex and is replaced by NOLA1/GAR1 to yield mature H/ACA snoRNPs complex. Probably competes with NOLA1/GAR1 for binding with DKC1/NOLA4.
Specifications
| NAF1 | |
| Polyclonal | |
| Unconjugated | |
| NAF1 | |
| Gm174; H/ACA ribonucleoprotein complex non-core subunit NAF1; hNAF1; NAF1; nuclear assembly factor 1 homolog; nuclear assembly factor 1 homolog (S. cerevisiae); nuclear assembly factor 1 ribonucleoprotein; RGD1306802 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96HR8 | |
| NAF1 | |
| Recombinant protein corresponding to Human NAF1. Recombinant protein control fragment (Product #RP-96681). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction