missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559836
This item is not returnable.
View return policy
Description
Immunogen sequence: KPFPELREHF QSYDLDHMEK KDHSHTPWIV IIAKYLAQWY SETNGRIPKT YKEKEDFRDL IRQGILKNEN GAPEDEENFE EAIKNVNTAL NTTQIPSSIE DIFNDDR Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| NAE1 | |
| Polyclonal | |
| Unconjugated | |
| NAE1 | |
| 59kDa; A-116A10.1; amyloid beta precursor protein binding protein 1, 59kDa; amyloid beta precursor protein-binding protein 1, 59 kDa; amyloid beta precursor protein-binding protein 1, 59kD; amyloid protein-binding protein 1; Appbp1; APP-BP1; HPP1; Nae1; NEDD8 activating enzyme E1 subunit 1; NEDD8-activating enzyme E1 regulatory subunit; NEDD8-activating enzyme E1 subunit; protooncogene protein 1; proto-oncogene protein 1; ula-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 234664, 84019, 8883 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13564, Q8VBW6, Q9Z1A5 | |
| NAE1 | |
| Recombinant protein corresponding to Human NAE1. Recombinant protein control fragment (Product #RP-98683). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction