missing translation for 'onlineSavingsMsg'
Learn More

NADPH tetrasodium salt, MedChemExpress

Product Code. p-200098690
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16458926

Brand: Medchem Express HYF0003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NADPH tetrasodium salt functions as an important cofactor in a variety of metabolic and biosynthetic pathways.

NADPH tetrasodium salt plays a vital role in the biosynthesis of drugs, chiral alcohols, fatty acids and biopolymers, while also being required for lipid biosynthesis, biomass formation, and cell replication. The demand for NADPH tetrasodium salt is particularly high in proliferating cancer cells, where it acts as a cofactor for the synthesis of nucleotides, proteins, and fatty acids. NADPH tetrasodium salt is also essential for the neutralization of the dangerously high levels of reactive oxygen species (ROS) generated by increased metabolic activity. NADPH tetrasodium salt is an endogenous inhibitor of ferroptosis.

Specifications

Chemical Name or Material NADPH tetrasodium salt
CAS 2646-71-1
Color White
Purity Grade Notes Research
Molecular Formula C21H26N7Na4O17P3
Quantity 100 mg
Solubility Information H2O : ≥ 35 mg/mL (42.00 mM) ∣DMSO : 10 mg/mL (12.00 mM; ultrasonic and warming and adjust pH to 3 with HCl and heat to 60°C)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O[Na])(O[Na])=O)[C@H](O)[C@@H](COP(OP(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C=C(C(N)=O)CC=C5)O4)(O[Na])=O)(O[Na])=O)O3)C=N2)=NC=N1
Molecular Weight (g/mol) 833.35
Formula Weight 833.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.