missing translation for 'onlineSavingsMsg'
Learn More

NADPH tetrasodium salt, MedChemExpress

Product Code. 16454697
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454697 50 mg 50mg
16458926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16454697 Supplier Medchem Express Supplier No. HYF000350mg
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NADPH tetrasodium salt functions as an important cofactor in a variety of metabolic and biosynthetic pathways.

NADPH tetrasodium salt plays a vital role in the biosynthesis of drugs, chiral alcohols, fatty acids and biopolymers, while also being required for lipid biosynthesis, biomass formation, and cell replication. The demand for NADPH tetrasodium salt is particularly high in proliferating cancer cells, where it acts as a cofactor for the synthesis of nucleotides, proteins, and fatty acids. NADPH tetrasodium salt is also essential for the neutralization of the dangerously high levels of reactive oxygen species (ROS) generated by increased metabolic activity. NADPH tetrasodium salt is an endogenous inhibitor of ferroptosis.

Specifications

Chemical Name or Material NADPH tetrasodium salt
CAS 2646-71-1
Color White
Purity Grade Notes Research
Molecular Formula C21H26N7Na4O17P3
Quantity 50 mg
Solubility Information H2O : ≥ 35 mg/mL (42.00 mM) ∣DMSO : 10 mg/mL (12.00 mM; ultrasonic and warming and adjust pH to 3 with HCl and heat to 60°C)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O[Na])(O[Na])=O)[C@H](O)[C@@H](COP(OP(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C=C(C(N)=O)CC=C5)O4)(O[Na])=O)(O[Na])=O)O3)C=N2)=NC=N1
Molecular Weight (g/mol) 833.35
Formula Weight 833.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.