missing translation for 'onlineSavingsMsg'
Learn More

NADPH tetracyclohexanamine, MedChemExpress

Product Code. 16468778
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468778 10 mg 10mg
16464697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468778 Supplier Medchem Express Supplier No. HYF0003A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NADPH tetracyclohexanamine is a ubiquitous cofactor and biological reducing agent.

NADPH tetracyclohexanamine is a ubiquitous cofactor and biological reducing agent.

Specifications

Chemical Name or Material NADPH tetracyclohexanamine
CAS 100929-71-3
Color White
Purity Grade Notes Research
Molecular Formula C45H82N11O17P3
Quantity 10 mg
Solubility Information H2O : ≥ 100 mg/mL (87.56 mM) ∣DMSO : ≥ 84 mg/mL (73.55 mM)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O)(O)=O)[C@H](O)[C@@H](COP(OP(O)(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C=C(C(N)=O)CC=C5)O4)=O)(O)=O)O3)C=N2)=NC=N1.NC6CCCCC6.NC7CCCCC7.NC8CCCCC8.NC9CCCCC9
Molecular Weight (g/mol) 1142.12
Formula Weight 1142.12
Percent Purity 96.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.