missing translation for 'onlineSavingsMsg'
Learn More

NADP sodium salt, MedChemExpress

Product Code. p-200100110
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16452326

Brand: Medchem Express HYF0002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NADP sodium salt (Sodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP sodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

NADP sodium salt (Sodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP sodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

Specifications

Chemical Name or Material NADP sodium salt
CAS 1184-16-3
Color White
Purity Grade Notes Research
Molecular Formula C21H27N7NaO17P3
Quantity 500 mg
Synonym Sodium NADP
Solubility Information H2O : 100 mg/mL (130.65 mM; Need ultrasonic) ∣DMSO : 3.57 mg/mL (4.66 mM; ultrasonic and warming and heat to 80°C)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O)(O)=O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H]([N+]5=CC=CC(C(N)=O)=C5)O4)=O)([O-])=O)O3)C=N2)=NC=N1.[Na+]
Molecular Weight (g/mol) 765.39
Formula Weight 765.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.