missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NADP disodium salt, MedChemExpress

Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
NADP disodium salt (Disodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP disodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).
Specifications
Specifications
Chemical Name or Material | NADP disodium salt |
CAS | 24292-60-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C21H28N7O17P3.2Na |
Quantity | 100 mg |
Synonym | Disodium NADP |
Solubility Information | H2O : 250 mg/mL (316.70 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (4.22 mM; ultrasonic and warming and heat to 80°C) |
SMILES | NC1=C2C(N([C@H]3[C@H](OP(O)(O)=O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H]([N+]5=CC=CC(C(N)=O)=C5)O4)=O)(O)=O)O3)C=N2)=NC=N1.[2 Na] |
Molecular Weight (g/mol) | 789.38 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction