missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NADH disodium salt, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
Description
NADH disodium salt (Disodium NADH) is an orally active reduced coenzyme. NADH disodium salt is a donor of ADP-ribose units in ADP-ribosylaton reactions and a precursor of cyclic ADP-ribose. NADH disodium salt plays a role as a regenerative electron donor in cellular energy metabolism, including glycolysis, β-oxidation and the tricarboxylic acid (TCA) cycle.
Specifications
Specifications
| Chemical Name or Material | NADH disodium salt |
| CAS | 606-68-8 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27N7Na2O14P2 |
| Quantity | 500 mg |
| Synonym | Disodium NADH |
| Solubility Information | H2O : ≥ 100 mg/mL (140.96 mM) ∣DMSO : 100 mg/mL (140.96 mM; Need ultrasonic) |
| SMILES | NC1=C2C(N([C@H]3[C@H](O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C=C(C(N)=O)CC=C5)O4)=O)([O-])=O)O3)C=N2)=NC=N1.[Na+].[Na+] |
| Molecular Weight (g/mol) | 709.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction