missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NADE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110029
This item is not returnable.
View return policy
Description
Immunogen sequence: MANIHQENEE MEQPMQNGEE DRPLGGGEGH QPAGNRRGQA RRLAPNFRWA IPNRQINDGM GGDGDDMEIF MEEMREIRRK LRELQLRNCL RILMGELSNH HDHHDEFCLM P.
May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death. May play an important role in the pathogenesis of neurogenetic diseases.
Specifications
| NADE | |
| Polyclonal | |
| Unconjugated | |
| BEX3 | |
| AL033356; Bex; Bex3; brain expressed X-linked 3; brain-expressed X-linked protein 3; Brain-expressed X-linked protein 3 homolog; DXS6984E; DXWsu67e; Gcap27; granule cell antiserum positive 27; HGR74; Nade; nerve growth factor receptor (TNFRSF16) associated protein 1; nerve growth factor receptor associated protein 1; nerve growth factor receptor-associated protein 1; Ngfrap1; ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated cell death executor; protein BEX3; rBex3; RP13-349O20.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12070, 27018 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q00994, Q9WTZ9 | |
| BEX3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-111 of human NGFRAP1 (NP_996798.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction