missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Detection method: Colorimetric/Fluorometric
- Assay time: 2 hr 30 min
- Results type: Quantitative
- Sample types: Tissue lysate and cell lysate
- Cited in: 19 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Western Blot Kit |
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 250 Tests |
| Substrate Properties | Chemical Substrate |
| Target Molecule | NAD, NADH |
| Detection Method | Colorimetric, Chromogenic, Chemiluminescence |
| Sample Type | Tissue Lysate, Cell Lysate |
| Quantity | 250 Tests |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?