missing translation for 'onlineSavingsMsg'
Learn More

NAD+, MedChemExpress

Product Code. 16422179
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422179 1 g 1g
16400938 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422179 Supplier Medchem Express Supplier No. HYB0445
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.

NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.

Specifications

Chemical Name or Material NAD+
CAS 53-84-9
Color White
Purity Grade Notes Research
Molecular Formula C21H27N7O14P2
Quantity 1 g
Synonym β-DPN β-NAD β-Nicotinamide Adenine Dinucleotide
Solubility Information H2O : 41.67 mg/mL (62.81 mM; Need ultrasonic)
SMILES O[C@H]([C@@H]1O)[C@@H](O[C@@H]1COP(OP(OC[C@H]2O[C@@H]([N+]3=CC=CC(C(N)=O)=C3)[C@H](O)[C@@H]2O)([O-])=O)(O)=O)N4C(N=CN=C5N)=C5N=C4
Molecular Weight (g/mol) 663.43
Formula Weight 663.43
Percent Purity 98.24%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.